Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 4/ANGPTL4 Antibody (Kairos-1), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB172313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172313 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172313 Supplier Novus Biologicals Supplier No. NBP280039DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Kairos-1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Host Species Mouse
Immunogen Recombinant human Angiopoietin-like Protein 4/ANGPTL4.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51129
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.