Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 4/ANGPTL4 Rabbit anti-Human, Clone: S04-1B5, Novus Biologicals™
SDP

Catalog No. NB188346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188346 100 μg
NB188347 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188346 Supplier Novus Biologicals Supplier No. NBP314995100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Applications Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone S04-1B5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Host Species Rabbit
Immunogen Recombinant protein of human Angiopoietin-like Protein 4/ANGPTL4 (Uniprot#: Q9BY76)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51129
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.