Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 6/ANGPTL6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Angiopoietin-like Protein 6/ANGPTL6
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126227
SDP
View Documents
Novus Biologicals
NBP310663100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Angiopoietin-like Protein 6/ANGPTL6 Polyclonal specifically detects Angiopoietin-like Protein 6/ANGPTL6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Angiopoietin-like Protein 6/ANGPTL6
Western Blot
Unconjugated
Rabbit
Human
AGFAngiopoietin-related protein 5, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ARP5angiopoietin-related protein 6
The immunogen for Anti-Angiopoietin-like Protein 6/ANGPTL6 antibody is: synthetic peptide directed towards the C-terminal region of Human ANGL6 (NP_114123.2). Peptide sequence PESDHYRLRLGQYHGDAGDSLSWHNDKPFSTVDRDRDSYSGNCALYQRGG
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
83854
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.