Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like protein 8/Betatrophin Antibody (OTI1B12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP272046A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272046A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272046A70 Supplier Novus Biologicals Supplier No. NBP272046AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiopoietin-like protein 8/Betatrophin Monoclonal specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Angiopoietin-like protein 8/Betatrophin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B12
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599
Gene Symbols ANGPTL8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 22-198 of human Angiopoietin-like protein 8/Betatrophin (NP_061157) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55908.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.