Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin I Antibody (BGN/KA/4H), HRP, Novus Biologicals™
SDP

Catalog No. NB10062344H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062344H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062344H Supplier Novus Biologicals Supplier No. NB10062344H

Mouse Monoclonal Antibody

Angiotensin I Monoclonal specifically detects Angiotensin I in Human, Rat samples. It is validated for ELISA.

Specifications

Antigen Angiotensin I
Applications ELISA
Classification Monoclonal
Clone BGN/KA/4H
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Gene Alias alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I
Gene Symbols AGT
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 40-49 of the angiotensinogen precursor (Swiss-Prot entry P01019) conjugated to HMW carrier protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 183
Test Specificity NB100-62344 is specific for Angiotensin I and does not react with angiotensin II, III or angiotensinogen in ELISA. Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. Angiotensin I is important in the Renin-Angiotensin-Aldosterone System (RAAS). RAAS is responsible for the regulation of blood pressure and water retention in the body. Angiotensin I is produced from a reaction between the renal enzyme Renin and angiotensinogen. It is converted by Angiotensin Converting Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.