Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin II Antibody (KAA8), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340469 Supplier Novus Biologicals Supplier No. NBP320123MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Angiotensin II Monoclonal antibody specifically detects Angiotensin II in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen Angiotensin II
Applications Western Blot
Classification Monoclonal
Clone KAA8
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AGT, alpha-1 antiproteinase, antitrypsin, angiotensin I, ANHU, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8
Host Species Rabbit
Immunogen Angiotensin II
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.