Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin II/III Antibody (Ang II E7 (BGN/KA/4L)), Biotin, Novus Biologicals™
SDP

Catalog No. NB10062346B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062346B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062346B Supplier Novus Biologicals Supplier No. NB10062346B

Mouse Monoclonal Antibody

Angiotensin II/III Monoclonal specifically detects Angiotensin II/III in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Angiotensin II/III
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ang II E7 (BGN/KA/4L)
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias AGT angiotensinogen (serpin peptidase inhibitor, clade A, member 8), angiotensin I, angiotensin II, ANHU, SERPINA8
Gene Symbols AGT
Host Species Mouse
Immunogen Peptide sequence corresponding to angiotensin II, conjugated to a proprietary molecular weight carrier. Peptide sequence: DRVYIHPF
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 183
Test Specificity This product reacts with Angiotensin II and III. Negligible reactivity was observed with Angiotensin I and Angiotensinogen.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.