Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANGPTL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118951
This item is not returnable.
View return policy
Description
ANGPTL6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
Specifications
ANGPTL6 | |
Polyclonal | |
Unconjugated | |
ANGPTL6 | |
6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70726 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8R0Z6 | |
ANGPTL6 | |
Recombinant protein Angiopoietin Like Protein 6. The antigen corresponds to amino acid range 238-457 of the target protein. | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction