Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANGPTL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $577.00
Specifications
Antigen | ANGPTL6 |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118951
|
Invitrogen™
PA5118951 |
200 μL |
Each for $577.00
|
|
|||||
PIPA5122240
|
Invitrogen™
PA5122240 |
20 μL |
Each for $237.00
|
|
|||||
Description
ANGPTL6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.Specifications
ANGPTL6 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
ANGPTL6 | |
6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
ANGPTL6 | |
Primary | |
Antigen affinity chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8R0Z6 | |
70726 | |
Recombinant protein Angiopoietin Like Protein 6. The antigen corresponds to amino acid range 238-457 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title