Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANGPTL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122240
This item is not returnable.
View return policy
Description
ANGPTL6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
Specifications
ANGPTL6 | |
Polyclonal | |
Unconjugated | |
ANGPTL6 | |
6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70726 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8R0Z6 | |
ANGPTL6 | |
Recombinant protein Angiopoietin Like Protein 6. The antigen corresponds to amino acid range 238-457 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction