Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ani 9
GREENER_CHOICE

Catalog No. p-7121035
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective TMEM16A (Anoctamin-1; ANO1) blocker

Ani 9 is a potent and selective TMEM16A (Anoctamin-1; ANO1) blocker (IC50 = 77 nM). Displays no effect on ANO2 activity (at 10 μM). Also exhibits no effect on intracellular calcium signaling or CFTR and ENaC channel activity (at 30 μM).

Specifications

Chemical Name or Material Ani 9
CAS 356102-14-2
Synonym 2-(4-Chloro-2-methylphenoxy)-N'-(2-methoxybenzylidene)acetohydrazide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C17H17ClN2O3
Formula Weight 332.78
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.