Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANIB1 Antibody (5H3), Novus Biologicals™
SDP

Catalog No. NBP237564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237564 Supplier Novus Biologicals Supplier No. NBP237564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANIB1 Monoclonal specifically detects ANIB1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ANIB1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5H3
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias AIB, AIB1, Aneurysm, Intracranial Berry 1
Host Species Mouse
Immunogen Purified recombinant fragment of human AIB1 expressed in E. Coli.
Molecular Weight of Antigen 155 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116833
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.