Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANIB1 Antibody (5H3), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | ANIB1 |
---|---|
Clone | 5H3 |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
ANIB1 Monoclonal specifically detects ANIB1 in Human samples. It is validated for Western Blot, ELISA.Specifications
ANIB1 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
116833 | |
IgG1 | |
Unpurified |
5H3 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
AIB, AIB1, Aneurysm, Intracranial Berry 1 | |
Purified recombinant fragment of human AIB1 expressed in E. Coli. | |
Primary | |
155 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title