Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Aniline Blue, sodium salt, pure, water soluble, certified
Click to view available options
Quantity:
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorption Ratio | 0.96 to 1.09 |
| Concentration | 12.0mL of 0.1N TiCl3/g min. |
| Infrared Spectrum | Authentic |
| Lambda Max | 594 to 610nm |
| Formula Weight | 737.71 |
| Percent Purity | 12.0 mL of 0.1 N TiCl3/g min. |
| Grade | Pure |
| Packaging | Glass bottle |
| Vapor Pressure | Vapor Pressure: Negligible |
| Color | Brown/Purple |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: WARNING! |
| Hazard Statement | GHS H Statement: Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. NFPA Health:2 Flammability:0 Instability:0 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction