Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Johannes Lieder Animal, Human and Plant Cytology
Shop All Johannes Lieder GmbH & Co. KG Products

Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 79600 Animal, Human and Plant Cytology
- Special Set comprising 25 prepared microscope slides of best quality.
- Simple animal cells in sec. of salamander liver showing nuclei, cell membranes and cytoplasm.
- For general study of the animal cell
- Mitotic stages in smear of red bone marrow of mammal
- Meiotic (maturation) stages in testis of mouse, sec. iron hematoxyline stained after Heidenhain
- Meiotic (maturation) stages in sec. through testis of salamander, selected material showing large structures
- Barr bodies (human sex chromatin) in smear from female squamous epithelium
- Mitochondria in thin sec. of kidney or liver, specially prepared and stained
- Golgi apparatus in sec. of spinal ganglion or other organ
- Pigment cells in skin
- Storage of glycogen in liver cells, sec. stained with carmine after Best or PAS reaction
- Storage of fat in cells of costal cartilage, sec. stained with Sudan
- Secretion of fat in mammary gland, section stained with Osmic acid
- Phagocytosis in Kupffer's star cells of the liver, sec. of mammalian liver injected with trypan blue
- Giant chromosomes in smear of the salivary gland of Chironomus larva, carefully fixed and stained
- Ascaris megalocephala embryology. Sec. of uteri showing entrance and modification of sperm in ova
- Ascaris megalocephala embryology. Sec. of uteri showing maturation stages (meiosis). Polar bodies can be seen.
- Ascaris megalocephala embryology. Sec. of uteri showing ova with male and female pronuclei
- Ascaris megalocephala embryology. Sec. of uteri showing early cleavage stages (mitosis)
- Ascaris megalocephala embryology. Sec. of uteri showing later cleavage stages (mitosis)
- Mitosis, l.s. from Allium root tips showing all stages of plant mitosis carefully stained with iron hematoxyline after Heidenhain
- DNA and RNA, thin l.s. from Allium root tips, specially fixed and stained with methylgreen and pyronine to show DNA and RNA in different colors
- Mitochondria, thin l.s. of Allium root tips specially fixed and stained to show the mitochondria clearly
- Meiosis, t.s. of Lilium anthers showing different stages of meiotic divisions
- Aleurone grains, sec. of Ricinus endosperm
- Inulin crystals, t.s. of tuber of Dahlia
- Chloroplasts, w.m. of leaf of Elodea or Spinacea showing detail of large chloroplasts
Specifications
Specifications
Classification | Cell Biology |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction