Learn More
Anisole, 99%
Description
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3',6'-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3′,6′-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
Solubility
Miscible with methanol and acetone. Immiscible with water.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -37°C |
| Density | 0.994 |
| Boiling Point | 153°C to 154°C |
| Flash Point | 43°C (109°F) |
| Odor | Ether-like |
| Refractive Index | 1.517 |
| Quantity | 500 g |
| UN Number | UN2222 |
| Beilstein | 506892 |
| Merck Index | 14,669 |
| Show More |
Safety and Handling
| Hazard Category | H226-H336 |
| Hazard Statement | GHS H Statement H226-H332-H315-H319-H335 Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ANISOLE |
| EINECSNumber | 202-876-1 |
| RTECSNumber | BZ8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.