Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Anisomycin

Click to view available options
Quantity:
10 mg
50 mg
Description
Anisomycin is a protein synthesis inhibitor (blocks translation). Potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. Acts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 265.31 |
Product Type | Anisomycin |
Purity | >98% |
Molecular Weight (g/mol) | 265.31 |
Chemical Name or Material | Anisomycin |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction