Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKHD1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290920 Supplier Novus Biologicals Supplier No. NBP178211PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKHD1 Polyclonal antibody specifically detects ANKHD1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ANKHD1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ankyrin repeat and KH domain containing 1, FLJ10042, FLJ11979, FLJ14127, hMASK, KIAA1085single KH-domain homolog, MASKFLJ20288, Multiple ankyrin repeats single KH domain
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 25 and 75 of human Ankyrin Repeat and KH Domain Containing 1 using the numbering given in entry NP_060217.1 (GeneID 54882).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 54882
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.