Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKK1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP128459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128459 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128459 Supplier Novus Biologicals Supplier No. NBP128459
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ANKK1 Polyclonal specifically detects ANKK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen ANKK1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5-10 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_848605.1
Gene Alias ankyrin repeat and kinase domain containing 1, ankyrin repeat and protein kinase domain-containing protein 1, EC 2.7.11, EC 2.7.11.1, PKK2, Protein kinase PKK2, SGK288, Sugen kinase 288, X-kinasesgK288
Gene Symbols ANKK1
Host Species Goat
Immunogen Peptide with sequence C-SRQADPNLHEAEGKT corresponding to internal region according to NP_848605.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255239
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.