Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKMY2 Antibody (OTI1F11), PE, Novus Biologicals™
SDP

Catalog No. NB288401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288401 Supplier Novus Biologicals Supplier No. NBP272073PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANKMY2 Monoclonal antibody specifically detects ANKMY2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen ANKMY2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F11
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias ankyrin repeat and MYND domain containing 2, ankyrin repeat and MYND domain-containing protein 2, DKFZP564O043, ZMYND20
Host Species Mouse
Immunogen Full length human recombinant protein of human ANKMY2(NP_064715) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57037
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.