Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD27 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB184652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184652 Supplier Novus Biologicals Supplier No. NBP282072AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen ANKRD27
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein
Host Species Rabbit
Immunogen ANKRD27 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human ANKRD27. The immunogen is located within amino acids 50 - 100 of ANKRD27. Amino Acid Squence: HFQTLNGKDVFIQGNR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84079
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.