Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD27 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291275 Supplier Novus Biologicals Supplier No. NBP150030PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ANKRD27
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1000 and 1050 of human Vps9 Domain and Ankyrin-Repeat-Containing Protein using the numbering given in entry NP_115515.2 (GeneID 84079).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84079
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.