Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD28 Antibody, Novus Biologicals™
SDP

Catalog No. NB10041419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041419 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041419 Supplier Novus Biologicals Supplier No. NB10041419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD28 Polyclonal specifically detects ANKRD28 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ANKRD28
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500-1:2000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O15084
Gene Alias ankyrin repeat domain 28, Ankyrin repeat domain-containing protein 28, KIAA0379PITKPP6-ARS-Aphosphatase interactor targeting K protein, Phosphatase interactor targeting protein hnRNP K, protein phosphatase 6 ankyrin repeat subunit A, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-A
Gene Symbols ANKRD28
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 975 and 1025 of human ankyrin repeat domain 28 (phosphatase interactor targeting K protein) using the numbering given in entry NP_056014.2 (GeneID 23243).
Molecular Weight of Antigen 113 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23243
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Rat (100%), Bovine (100%), Horse (100%), Rabbit (100%), Pig (100%), Panda (100%), Oragutan (100%), Rhesus Monkey (100%), Gorilla (100%), Chimpanzee (100%), White-tufted-ear marmoset (100%), Naked mole rat (100%), Little brown bat (100%), Northern white-cheeked gibbon (100%), and Small-eared galago (100%).
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.