Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD50 Antibody, Novus Biologicals™
SDP

Catalog No. NBP150031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP150031 Supplier Novus Biologicals Supplier No. NBP150031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD50 Polyclonal specifically detects ANKRD50 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ANKRD50
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias ankyrin repeat domain 50
Gene Symbols ANKRD50
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1200 and 1250 of human Ankyrin Repeat Domain 50 using the numbering given in entry NP_065070.1 (GeneID 57182).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57182
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.