Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD53 Antibody (OTI1E1), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272217PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272217PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272217PEC5 Supplier Novus Biologicals Supplier No. NBP272217PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANKRD53 Monoclonal specifically detects ANKRD53 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ANKRD53
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1E1
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ankyrin repeat domain 53, ankyrin repeat domain-containing protein 53, FLJ12056, FLJ36160
Gene Symbols ANKRD53
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-300 of human ANKRD53 (NP_079209) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79998.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.