Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131621 Supplier Novus Biologicals Supplier No. NBP131621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD7 Polyclonal specifically detects ANKRD7 in Human samples. It is validated for Western Blot.

Specifications

Antigen ANKRD7
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q92527
Gene Alias ankyrin repeat domain 7, ankyrin repeat domain-containing protein 7, testis-specific ankyrin motif containing protein, Testis-specific protein TSA806, TSA806
Gene Symbols ANKRD7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKRD7. The exact sequence is proprietary.
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56311
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.