Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15690020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15690020 20 μL
NBP156900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15690020 Supplier Novus Biologicals Supplier No. NBP15690020UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ANKRD9 Polyclonal specifically detects ANKRD9 in Human samples. It is validated for Western Blot.

Specifications

Antigen ANKRD9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96BM1
Gene Alias ankyrin repeat domain 9, ankyrin repeat domain-containing protein 9, MGC21990
Gene Symbols ANKRD9
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ANKRD9(ankyrin repeat domain 9) The peptide sequence was selected from the middle region of ANKRD9 (NP_689539). Peptide sequence RARVLDRRGCSRVEGGGTSLHVACELARPECLFLLLGHGASPGLRDGGGL.
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122416
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.