Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10061625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061625 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061625 Supplier Novus Biologicals Supplier No. NB10061625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKS3 Polyclonal specifically detects ANKS3 in Canine, Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ANKS3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q6ZW76
Gene Alias ankyrin repeat and SAM domain-containing protein 3, ankyrin repeat and sterile alpha motif domain containing 3, FLJ32767, KIAA1977FLJ32345
Gene Symbols ANKS3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 606 and 656 of human ankyrin repeat and sterile alpha motif domain containing 3 using the numbering given in entry NP_597707.1 (GeneID 124401).
Molecular Weight of Antigen 72 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124401
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee.
Target Species Canine, Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.