Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin 3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB360343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360343 Supplier Novus Biologicals Supplier No. NBP305528MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ankyrin 3 Monoclonal antibody specifically detects Ankyrin 3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Ankyrin 3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier
Host Species Mouse
Immunogen The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 288
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.