Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin 3 Antibody (S106-20), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP259310C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259310C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259310C Supplier Novus Biologicals Supplier No. NBP259310C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ankyrin 3 Monoclonal specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ankyrin 3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S106-20
Conjugate DyLight 650
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier
Gene Symbols ANK3
Host Species Mouse
Immunogen Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of Ankyrin repeats
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 288
Test Specificity Detects >200kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.