Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin Brain Antibody (S105-13), HRP, Novus Biologicals™
SDP

Catalog No. NBP259374H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259374H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259374H Supplier Novus Biologicals Supplier No. NBP259374H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ankyrin Brain Monoclonal specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Ankyrin Brain
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone S105-13
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin
Gene Symbols ANK2
Host Species Mouse
Immunogen Synthetic peptide amino acids 203-496 of human Ankyrin-B
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 287
Test Specificity Detects 200kDa. No cross reactivity against Ankyrin-G.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.