Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin Repeat Domain 33B Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB149966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB149966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB149966 Supplier Novus Biologicals Supplier No. NBP298013AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ankyrin Repeat Domain 33B Polyclonal specifically detects Ankyrin Repeat Domain 33B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Ankyrin Repeat Domain 33B
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ANKRD33B, Ankyrin Repeat Domain-Containing Protein 33B, Ankyrin Repeat Domain-Containing Protein LOC651746
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Ankyrin Repeat Domain 33B fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 651746
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.