Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin repeat domain 39 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215402 Supplier Novus Biologicals Supplier No. NBP215402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ankyrin repeat domain 39 Polyclonal specifically detects Ankyrin repeat domain 39 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ankyrin repeat domain 39
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q53RE8
Gene Alias ankyrin repeat domain 39, ankyrin repeat domain-containing protein 39, MGC41816
Gene Symbols ANKRD39
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKRD39. The exact sequence is proprietary.
Molecular Weight of Antigen 20 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51239
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (92%), Rat (91%), Bovine (92%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.