Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Anle138b

Click to view available options
Quantity:
5 mg
25 mg
Description
Anle138b is a oligomeric aggregation inhibitor that inhibits formation of pathological prion protein (PrPSc) and α-synuclein aggregates in vitro. Anle138b prevents accumulation of PrPSc in brain and spleen, and prolongs survival of prion-infected mice. Anle138b exhibits neuroprotective effects in a Parkinson's disease mouse model and improves motor performance in an α-synucleinopathy model. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Quantity | 5 mg |
| Molecular Formula | C16H11BrN2O2 |
| Formula Weight | 343.18 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction