Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A1 Antibody (ANXA1/3869R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB379013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379013 Supplier Novus Biologicals Supplier No. NBP308709JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Annexin A1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ANXA1/3869R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Host Species Rabbit
Immunogen Recombinant full-length human Annexin A1 protein (Uniprot: P04083)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 301
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.