Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A10 Antibody (OTI4D8), FITC, Novus Biologicals™
SDP

Catalog No. NBP271683F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271683F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271683F Supplier Novus Biologicals Supplier No. NBP271683F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A10 Monoclonal specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Annexin A10
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4D8
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10
Gene Symbols ANXA10
Host Species Mouse
Immunogen Full length recombinant protein of human Annexin A10 (NP_009124) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11199.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.