Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A11 Antibody (OTI1C6), FITC, Novus Biologicals™
SDP

Catalog No. NBP270182F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270182F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270182F Supplier Novus Biologicals Supplier No. NBP270182F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A11 Monoclonal specifically detects Annexin A11 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Annexin A11
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1C6
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias annexin A11, Annexin-11, ANX1156-kD, CAP50, CAP-50
Gene Symbols ANXA11
Host Species Mouse
Immunogen Full length recombinant protein of human Annexin A11 (NP_001148) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 311.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.