Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A2 Antibody (ANEX 3D5/4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB1002724FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002724FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002724FR Supplier Novus Biologicals Supplier No. NB1002724FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A2 Monoclonal specifically detects Annexin A2 in Human samples. It is validated for ELISA, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Annexin A2
Applications ELISA, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ANEX 3D5/4
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I
Gene Symbols ANXA2
Host Species Mouse
Immunogen Mixture of native membrane proteins from human bone tissues which would include whole native annexin proteins
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 302
Test Specificity ANEX 3D5/4 detects native protein. Recommended concentration range: 1-10 ug/ml.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.