Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A4 Antibody (SAIC-14C-10F12), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB362916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362916 Supplier Novus Biologicals Supplier No. NBP320093MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Annexin A4 Monoclonal antibody specifically detects Annexin A4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry

Specifications

Antigen Annexin A4
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Mass Spectrometry
Classification Monoclonal
Clone SAIC-14C-10F12
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Formulation 50mM Sodium Borate
Gene Alias annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4Carbohydrate-binding protein p33/p41, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36
Host Species Rabbit
Immunogen Peptide DEGNYLDDALVR derived from Annexin A4 conjugated to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 307
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.