Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A7 Antibody (OTI1C3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270181FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270181FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270181FR Supplier Novus Biologicals Supplier No. NBP270181FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A7 Monoclonal specifically detects Annexin A7 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Annexin A7
Applications Western Blot
Classification Monoclonal
Clone OTI1C3
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias annexin A7, ANX7, ANXA7, SNX, SYNEXIN
Gene Symbols ANXA7
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 105-411 of human Annexin A7 (NP_004025) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 310.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.