Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A8/ANXA8 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB041765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041765 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041765 Supplier Novus Biologicals Supplier No. NBP297052B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Annexin A8/ANXA8 Polyclonal specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Annexin A8/ANXA8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Annexin A8/ANXA8 (AAH73755.1; Met1-Pro327).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 653145
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.