Learn More
Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kits

Description
Includes
- Cat. No. 88-8005-72:
- 10X Binding Buffer
- Annexin V FITC : 5μL/test, 50 tests Protect from light.
- Propidium Iodide Staining Solution : 5μL/test, 100 tests Protect from light.
- Cat. No. 88-8005-74:
- 10X Binding Buffer
- Annexin V FITC : 5μL/test, 200 tests Protect from light.
- Propidium Iodide Staining Solution : 5μL/test, 2x100 tests Protect from light.
Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.
In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).
Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

Specifications
Specifications
Description | eBioscience Annexin V Apoptosis Detection Kit FITC |
Quantity | 50 Tests |
Format | Kit |
Kit Contents | Contains: • Annexin V Binding Buffer (10X), 1 bottle (30 mL) • Annexin V-FITC conjugate, 1 vial (0.25 mL) • Propidium Iodide Staining Solution, 1 vial (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Content And Storage | Handling Conditions: Store at 2-8°C. Avoid temperature fluctuations. |
Excitation/Emission | 488, 535/520, 617 |
Product Line | eBioscience |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.