Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Annexin V Apoptosis Kit, Fluorescein
Shop All R&D Systems Products

Click to view available options
Quantity:
125 Tests
250 Tests
Description
- Annexin V is a member of a calcium and phospholipid binding family of proteins with vascular anticoagulant activity. It is also known as PP4, PAP I, CPB-I, CaBP33, VACa, anchorin CII, lipocortin-V, endonexin II, and thromboplastin inhibitor
- Facilitates rapid fluorometric quantitation of apoptotic cell
- Allows the use of streptavidinconjugated fluorophores other than FITC for detection
Applications: Flow cytometry
Conjugate: Fluorescein
Sample type: Unfixed cells
Specifications
Specifications
| Quantity | 125 Tests |
| Product Type | Apoptosis detection kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Protect from light. Do not use past expiration date. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction