Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V-FITC

Catalog No. p-4340663 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests
200 Tests
25 Tests

Catalog No. BDB556419

Supplier: BD 556419

Only null left
Add to Cart
Add to Cart

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.

  • Format: FITC

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Tests
Sample Volume 5 μL
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Conjugate FITC
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.