Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AnnexinA10 Rabbit anti-Human, Clone: SC06-02, Novus Biologicals™
SDP

Catalog No. NBP267600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267600 Supplier Novus Biologicals Supplier No. NBP267600

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

Annexin A10 Monoclonal specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AnnexinA10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SC06-02
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10
Gene Symbols ANXA10
Host Species Rabbit
Immunogen Recombinant protein within Human Annexin A10 aa 30-220/ 324. (SwissProt: Q9UJ72 Human; SwissProt: Q9QZ10 Mouse; Unigene:101584 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11199
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.