Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO6/TMEM16F Mouse anti-Human, Clone: N429/19, MilliporeSigma™

Mouse Monoclonal Antibody

$182.31 - $463.74

Specifications

Antigen ANO6/TMEM16F
Clone N429/19
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN186625
View Documents
MilliporeSigma
MABN186625UL
25 μL
Each for $182.31
Only null left
Add to Cart
 
MABN1866100
View Documents
MilliporeSigma
MABN1866100UL
100 μL
Each for $463.74
Only null left
Add to Cart
 
Description

Description

ANO6/TMEM16F Monoclonal antibody specifically detects ANO6/TMEM16F in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

ANO6/TMEM16F
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2b κ
Protein G purified
N429/19
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
Recombinant fragment corresponding to the first 294 amino acids from the N-terminal, cytoplasmic domain of human Anoctamin-6 (ANO6).
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.