Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANP32A Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB098933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098933 Supplier Novus Biologicals Supplier No. NBP299913UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANP32A Polyclonal specifically detects ANP32A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ANP32A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member A, acidic leucine-rich nuclear phosphoprotein 32 family member A, Acidic nuclear phosphoprotein pp32, cerebellar leucine rich acidic nuclear protein, I1PP2A, inhibitor-1 of protein phosphatase-2A, LANPPutative HLA-DR-associated protein I, Leucine-rich acidic nuclear protein, MAPMC15orf1, Mapmodulin, PHAP1MGC150373, PHAPIMGC119787, Potent heat-stable protein phosphatase 2A inhibitor I1PP2A, PP32, putative human HLA class II associated protein I
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ANP32A (NP_006296.1; Glu2-Lys238).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8125
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.