Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANP32B Antibody, Novus Biologicals™
SDP

Catalog No. NB1001199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001199 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001199 Supplier Novus Biologicals Supplier No. NB1001199
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

ANP32B Polyclonal specifically detects ANP32B in Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA, Western Blot.

Specifications

Antigen ANP32B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 3-5 ug/ml, Peptide ELISA Detection limit 1:16000, Knockdown Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006392
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member B, acidic leucine-rich nuclear phosphoprotein 32 family member B, Acidic protein rich in leucines, APRILSilver-stainable protein SSP29, Pal31, PHAPI2SSP29, Putative HLA-DR-associated protein I-2
Gene Symbols ANP32B
Host Species Goat
Immunogen Peptide with sequence C-KRKRETDDEGEDD corresponding to C-Terminus according to NP_006392.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10541
Test Specificity This antibody does NOT recognize theTNF family member also known as APRIL - Gene ID number 8741. However this antibody may cross-react with ANP32A (GeneID 8125).
Target Species Human, Mouse, Rat, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.