Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ANR 94
GREENER_CHOICE

Catalog No. 393710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-371-0 10 mg
39-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-371-0 Supplier Tocris Bioscience™ Supplier No. 3937/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adenosine A2A antagonist

Adenosine A2A receptor (AA2AR) antagonist (Ki values are 643 and 46 nM for rat and human AA2ARs respectively). Most active AA2AR antagonist for human receptors. Displays activity in the treatment of Parkinson's disease in vivo; improves parkinsonian motor deficits and tremors. Exhibits neuroprotective and anti-inflammatory effects.

Chemical Identifiers

CAS 634924-89-3
Molecular Formula C9H13N5O

Specifications

Quantity 10 mg
Formula Weight 207.23
Purity >99%
Molecular Weight (g/mol) 207.237
Chemical Name or Material ANR 94
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.