Learn More
Description
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
- Provide flexibility and tactility
- Made from an exceptionally thin chemical-resistant, five-layer laminated film
- Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
- Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
- Hand specific
- Broad spectrum chemical resistance
- Wing-thumb for added wearer comfort
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 0 |
| Certifications/Compliance (Europe) | CE 0493, EN420:2003 + A1:2009; Category lll |
| Applications | Industrial |
| Color | White |
| Hand | Specific |
| Latex Free | Yes |
| Material | Laminated Film |
| Product Type | Chemical-resistant Gloves |
| Size | 9 |
| Thickness (English) | 2.5 mil |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.